Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KIAA0922 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524886

Catalog No. PIPA524886

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

KIAA0922
Polyclonal
PBS with 0.09% sodium azide
A2VDJ0
TMEM131L, transmembrane protein 131-like
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
23240
Flow Cytometry, Western Blot
Unconjugated
TMEM131L
Liquid
TMEM131L
KLH conjugated synthetic peptide between 1266-1295 amino acids from the C-terminal region of human T131L
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit