Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-T7 Epitope Tag, Agarose, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP140393

Catalog No. NBP140393


Add to cart

Description

Description

T7 Epitope Tag Polyclonal antibody specifically detects T7 Epitope Tag in Non-species specific samples. It is validated for Immunoprecipitation.
Specifications

Specifications

T7 Epitope Tag
Agarose
PBS with 0.09% Sodium Azide
MASMTGGQQMG epitope tag, MASMTGGQQMG tag, T7 epitope tag
Rabbits were immunized with T7 cleavage site (MASMTGGQQMG) conjugated to KLH. Antibody was isolated by affinity chromatography using the peptide immobilized on solid support. Rabbit anti- human T7 affinity purified antibodies were coupled to peptide immobilized on solid support.
Immunogen affinity purified
RUO
Store at 4C. Do not freeze.
Polyclonal
Immunoprecipitation
Immunoprecipitation 15 -25
Affinity Purified
Rabbit
IgG
0.1 mg
Epitope Tags, T7 Epitope Tags
Primary
Non-species specific
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only