Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
T7 Tag Polyclonal Antibody, HRP, Invitrogen™
Goat Polyclonal Antibody
Supplier: Thermo Scientific PA131449
Description
PA1-31449 detects T7 tag from tagged samples.
T7 Tag Polyclonal antibody specifically detects T7 Tag in Tag samples. It is validated for ELISA, Immunocytochemistry, Immunohistochemistry, Western Blot.Specifications
T7 Tag | |
Polyclonal | |
HRP | |
MASMTGGQQMG; T7 cleavage | |
synthetic peptide, MASMTGGQQMG (T7), conjugated to KLH. | |
100 μg | |
Primary | |
Antibody | |
IgG |
ELISA, Immunocytochemistry, Immunohistochemistry, Western Blot | |
1 mg/mL | |
PBS with 0.2% BSA and 0.05% Pro-Clean 400; pH 7.2 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid | |
Tag |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction