Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

T7 Tag Goat anti-Tag, Polyclonal, Invitrogen™

Goat Polyclonal Antibody

Manufacturer:  Invitrogen PA129983

Catalog No. PA129983


Add to cart

Description

Description

PA1-29983 detects T7 tag from tagged samples. PA1-29983 has been successfully used in immunocytochemistry, ELISA, and Western blot applications. The PA1-29983 immunogen is: Synthetic peptide: MASMTGGQQMG (T7) conjugated to KLH.

The T7 Tag is an 11 amino acid peptide derived from the bacteriophage T7 major capsid protein, gene 10. T7 cleavage site: MASMTGGQQMG. Epitope tags provide a method to localize gene products in a variety of cell types, study the topology of proteins and protein complexes, identify associated proteins, and characterize newly identified, low abundance or poorly immunogenic proteins when protein specific antibodies are not available.
Specifications

Specifications

T7 tag
Polyclonal
Unconjugated
Liquid
Goat
IgG
100 μg
4° C, do not freeze
Primary
Tag
ELISA, Immunocytochemistry, Immunofluorescence, Western Blot
1 mg/mL
PBS with 0.1% sodium azide; pH 7.2
MASMTGGQQMG; T7 cleavage
synthetic peptide MASMTGGQQMG (T7) conjugated to KLH.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit