Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TAAR1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA120781

Catalog No. PIPA120781


Add to cart

Description

Description

PA1-20781 detects TA1 in Human samples. PA1-20781 has been successfully used in Immunohistochemistry (Paraffin) procedures. PA1-20781 immunogen corresponds to Synthetic peptide conjugated to KLH (aa sequence is considered to be commercially sensitive)..

TA1 (Trace amine receptor 1) is a member of the G protein coupled receptor family. It is activated by endogenous trace amines as well as metabolites of the biogenic amine neurotransmitters. Trace amines are biogenic amines present in very low levels in mammalian tissues. Although some trace amines have clearly defined roles as neurotransmitters in invertebrates, the extent to which they function as true neurotransmitters in vertebrates has remained speculative.
Specifications

Specifications

TAAR1
Polyclonal
Unconjugated
TAAR1
Liquid
TAAR1
Synthetic peptide conjugated to KLH (aa sequence is considered to be commercially sensitive).
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunohistochemistry (Paraffin)
1 mg/mL
PBS with 0.01% sodium azide; pH 7.7
Q96RJ0
MGC126874, MGC138399, RP11-295F4.9, TA1, TAR1, TRAR1
Rabbit
IgG
25 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
134864
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit