Learn More
TAB1 Rabbit anti-Human, Mouse, Rhesus Macaque, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Manufacturer:  MilliporeSigma 061336
Catalog No. 06-133-6MI
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
This Anti-TAB1 Antibody is validated for use in western blotting for the detection of TAB1.
Specifications
Specifications
TAB1
Unconjugated
Affinity Purified
Rabbit
Affinity purified
2-8°C for 12 months
Polyclonal
Human, Murine, Rhesus
Western Blot
Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
TAB1; MAP3K7IP
Histidine-tagged recombinant protein corresponding to human TAB1.
100μg
Primary
NP_006107