Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TACC2 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA522119

Catalog No. PIPA522119


Add to cart

Description

Description

PA5-22119 targets TACC2 in IHC (P) and WB applications and shows reactivity with Human samples. The PA5-22119 immunogen is synthetic peptide contain a sequence corresponding to a region within amino acids 237 and 293 (O95359) of TACC2. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

Specifications

Specifications

TACC2
Polyclonal
Unconjugated
TACC2
Liquid
TACC2
Synthetic peptide contain a sequence corresponding to a region within amino acids 237 and 293 (O95359) of TACC2
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunohistochemistry (Paraffin), Western Blot
0.28 mg/mL
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
O95359
RP11-296H2.1, AZU-1, ECTACC, anti zuai-1, transforming acidic coiled-coil-containing protein 2
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
10579
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit