Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TADA3L Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA527591
Description
Recommended positive controls: TADA3L tansfected 293T cell. Predicted reactivity: Mouse (100%), Rat (100%), Xenopus laevis (90%), Pig (100%), Rhesus Monkey (100%), Chimpanzee (99%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
TADA3L Polyclonal antibody specifically detects TADA3L in Human samples. It is validated for Western Blot.Specifications
TADA3L | |
Polyclonal | |
Unconjugated | |
TADA3 | |
1110004B19Rik; ADA3; ADA3 homolog; ADA3-like protein; AI987856; alteration/deficiency in activation 3; fa03f04; FLJ20221; FLJ21329; hADA3; mADA3; NGG1; STAF54; Tada3; Tada3l; Transcriptional adapter 3; transcriptional adapter 3-like; transcriptional adaptor 3; transcriptional adaptor 3 (NGG1 homolog, yeast)-like; transcriptional adaptor 3-like isoform b; Unknown (protein for MGC:133915); wu:fa03f04; zgc:63596 | |
Rabbit | |
Protein A | |
RUO | |
10474 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
9.46 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
O75528 | |
TADA3 | |
Recombinant fragment corresponding to a region within amino acids 251 and 432 of Human TADA3L | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction