Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TAF(II)135 Mouse, Unlabeled, Clone: 22, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 612054

Catalog No. BDB612054


Add to cart

Description

Description

Tightly associated factors (TAFs) play an essential role in transcriptional activation through their interaction with a variety of activators. TAF[II]135 (a.k.a. TAF[II]130) is a human homologue of Drosophila TAF[II]110, the first TAF shown to have coactivator activity. TAF[II]135 contains multiple glutamine-rich regions, as well as a coactivator domain (CAD). The glutamine-rich regions of TAF[II]135 facilitate interaction with Sp1 and CREB, which leads to enhancement of both Sp1- and CREB-mediated transcription. In addition, TAF[II]135 can potentiate transcriptional stimulation by AF-2 of the retinoic acid, thyroid hormone, and vitamine D3 receptors. However TAF[II]135 does not interact with the AF-2s of the estrogen and retinoid X receptors. Interestingly, TAF[II]135 enhancement of CREB transcriptional activity may be disrupted by expanded polyglutamine (CAG) repeats found in at least eight different neurodegenerative disorders. Thus, TAF[II]135 may have important coactivator activities in many different transcription-regulating cell signaling pathways, and interference of TAF[II]135 activity by CAG repeats may be a common mechanism of specific types of neuropathologies.

Host Species: Mouse
Clone: 22
Isotype: IgG1
Species Reactivity [for Features Main]: Human
Immunogen: Human TAF[II]135 aa. 454-565

Immunofluorescence, Western Blotting

Specifications

Specifications

TAF(II)135
22
Unconjugated
Affinity Purified
Human TAF[II]135 aa. 454-565
50μg
Cell Biology
Primary
Human, Murine
Western Blot
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Murine
IgG1
RUO
Store undiluted at -20°C.
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit