Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TAF11 RNA polymerase II, TATA box binding protein (TBP)-associated factor, 28kDa, Mouse, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a full-length human TAF11 protein.

Manufacturer:  Abnova Corporation H00006882B01

Catalog No. 89-006-317


Add to cart

Description

Description

Initiation of transcription by RNA polymerase II requires the activities of more than 70 polypeptides. The protein that coordinates these activities is transcription factor IID (TFIID), which binds to the core promoter to position the polymerase properly, serves as the scaffold for assembly of the remainder of the transcription complex, and acts as a channel for regulatory signals. TFIID is composed of the TATA-binding protein (TBP) and a group of evolutionarily conserved proteins known as TBP-associated factors or TAFs. TAFs may participate in basal transcription, serve as coactivators, function in promoter recognition or modify general transcription factors (GTFs) to facilitate complex assembly and transcription initiation. This gene encodes a small subunit of TFIID that is present in all TFIID complexes and interacts with TBP. This subunit also interacts with another small subunit, TAF13, to form a heterodimer with a structure similar to the histone core structure. [provided by RefSeq

Sequence: MDDAHESPSDKGGETGESDETAAVPGDPGATDTDGIPEETDGDADVDLKEAAAEEGELESQDVSDLTTVEREDSSLLNPAAKKLKIDTKEKKEKKQKVDEDEIQKMQILVSSFSEEQLNRYEMYRRSAFPKAAIKRLIQSITGTSVSQNVVIAMSGISKVFVGEVVEEALDVCEKWGEMPPLQPKHMREAVRRLKSKGQIPNSKHKKIIFF
Specifications

Specifications

TAF11 RNA polymerase II, TATA box binding protein (TBP)-associated factor, 28kDa
Unconjugated
No additive
NM_005643.2
MGC:15243|PRO2134|TAF2I|TAFII28
Mouse
50 uL
Transcription Regulation
Y
Polyclonal
Human
Western Blot
Mouse polyclonal antibody raised against a full-length human TAF11 protein.
TAF11
Serum
TAF11
TAF11 (NP_005634.1, 1 a.a. ~ 211 a.a) full-length human protein.
RUO
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Primary
6882
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit