Learn More
TAF13 Mouse anti-Human, Clone: 2B3, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA520069
Catalog No. PIMA520069
Add to cart
Description
Description
Peptide Sequence: KRKRLFSKEL RCMMYGFGDD QNPYTESVDI LEDLVIEFIT EMTHKAMSIG RQGRVQVEDI VFLIRKDPRK FARVKDLLTM NEELKRARKA FDEANYGS
Initiation of transcription by RNA polymerase II requires the activities of more than 70 polypeptides. The protein that coordinates these activities is transcription factor IID (TFIID), which binds to the core promoter to position the polymerase properly, serves as the scaffold for assembly of the remainder of the transcription complex, and acts as a channel for regulatory signals. TFIID is composed of the TATA-binding protein (TBP) and a group of evolutionarily conserved proteins known as TBP-associated factors or TAFs. TAFs may participate in basal transcription, serve as coactivators, function in promoter recognition or modify general transcription factors (GTFs) to facilitate complex assembly and transcription initiation. This gene encodes a small subunit associated with a subset of TFIID complexes. This subunit interacts with TBP and with two other small subunits of TFIID, TAF10 and TAF11. There is a pseudogene located on chromosome 6.
Specifications
Specifications
TAF13
Monoclonal
0.2-1.0 mg/mL
PBS with no preservative; pH 7.4
Q15543
TAF13 RNA polymerase II, MGC22425, TAF2K, TAFII18, TAFII-18, TAF(II)18
Mouse
IgG2a, kappa
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
6884
ELISA, Western Blot
2B3
Unconjugated
TAF13
Liquid
TAF13
TAF13 (NP_005636.1, 27 a.a. ∽ 124 a.a) partial recombit protein with GST tag. MW of the GST tag alone is 26 KDa.
Affinity chromatography
RUO
Antibody
Monoclonal
Human