Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TAFA3 Mouse anti-Human, Clone: 460904, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA524180

Catalog No. PIMA524180


Add to cart

Description

Description

In direct ELISAs and Western blots, no cross-reactivity with Recombinant human TAFA1, 2, 4, or 5 is observed. Reconstitute at 0.5 mg/mL in sterile PBS.

Specifications

Specifications

TAFA3
Monoclonal
0.5 mg/mL
PBS with 5% trehalose and No Preservative
Q7Z5A8
TAFA-3, TAFA3, chemokine-like protein TAFA-3, protein FAM19A3
Mouse
IgG2B
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
284467
Flow Cytometry, Western Blot
460904
Unconjugated
FAM19A3
Lyophilized
FAM19A3
E. coli-derived recombit human TAFA3/FAM19A3 Ala31-Arg133
Protein A/G
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit