Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-tag-192, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner 29430002

Catalog No. 29-430-002


Add to cart

Description

Description

tag-192 Polyclonal antibody specifically detects tag-192 in C. elegans samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

tag-192
1 mg/mL
ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:30000
T04D1.4
Protein CHD-7
Rabbit
IgG
0.05 ml
modENCODE Antibodies
Primary
172079
C. elegans
ELISA, Immunocytochemistry, Immunofluorescence
Unconjugated
20mM Potassium Phosphate (pH 7.0) and 0.15M NaCl with No Preservative
Affinity Purified
chd-7
In vivo generated recombinant protein fragment
Immunogen affinity purified
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Polyclonal
Caenorhabditis elegans TAG192
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only