Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TAGAP Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA524442
Description
TAGAP Polyclonal antibody specifically detects TAGAP in Human samples. It is validated for Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot.Specifications
TAGAP | |
Polyclonal | |
Unconjugated | |
TAGAP | |
2610315E15Rik; ARHGAP47; FKSG15; IDDM21; T cell activation Rho GTPase activating protein; Tagap; TAGAP1; T-cell activation GTPase activating protein; T-cell activation GTPase-activating protein; T-cell activation GTPase-activating protein 1; T-cell activation Rho GTPase-activating protein; T-cell activation RhoGTPase activating protein | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
117289 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
PBS with 0.09% sodium azide | |
Q8N103 | |
TAGAP | |
KLH conjugated synthetic peptide between 103-129 amino acids from the central region of human TAGAP | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction