Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TAL1 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA512337
Description
TAL1 is implicated in the genesis of hemopoietic maligcies. It may play an important role in hemopoietic differentiation. It serves as a positive regulator of erythroid differentiation.Specifications
TAL1 | |
Polyclonal | |
TAL1 | |
T-cell acute lymphocytic leukemia protein 1; TAL-1; Stem cell protein; T-cell leukemia/lymphoma protein 5; TAL1; SCL; TCL5 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
6886 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
Unconjugated | |
P17542 | |
TAL1 | |
KLH conjugated synthetic peptide between 68-97 amino acids from human TAL1 | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title