Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Talin 2, Polyclonal, Invitrogen™

Chicken Polyclonal Antibody

Manufacturer:  Invitrogen PA129481

Catalog No. PIPA129481

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This gene encodes a protein related to talin 1, a cytoskeletal protein that plays a significant role in the assembly of actin filaments and in spreading and migration of various cell types, including fibroblasts and osteoclasts. This protein has a different pattern of expression compared to talin 1 but, like talin 1, is thought to associate with unique transmembrane receptors to form novel linkages between extracellular matrices and the actin cytoskeleton.

PA1-29481 detects Talin 2 from mouse, human samples.PA1-29481 has been successfully used in Western blot applications.The PA1-29481 immunogen is: Amino acids 1750-1950 of Talin 2.
Specifications

Specifications

Talin 2
Polyclonal
Unconjugated
TLN2
Purified
TLN2
Amino acids 1750-1950 of Talin 2.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Murine
Western Blot
1mg/mL
PBS with no preservative
Q9Y4G6, Q71LX4
DKFZp451B1011, DKFZp686I0976, DKFZp686K0979, ILWEQ, KIAA0320
Chicken
IgY
50μg
Store at -20°C
Primary
83660
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit