Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Talin Mouse anti-Human, Mouse, Rabbit, Clone: TA205, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma 05385
Description
Anti-Talin (human) Antibody, clone TA205 is an antibody against Talin (human) for use in immunoprecipitation, western blot, immunohistochemistry.Specifications
Talin | |
Monoclonal | |
Unconjugated | |
NM_006289 | |
Mouse | |
Protein A chromatography | |
Primary | |
−20°C for 24 months. | |
IgG1 |
Immunohistochemistry, Immunoprecipitation, Western Blot | |
TA205 | |
PBS, pH 7.4, 0.05% sodium azide | |
TLN1, KIAA1027, Talin-1., ILWEQ, TLN, talin | |
Human Talin | |
200 μg | |
Human, Mouse, Rabbit | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only