Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Talin Mouse anti-Human, Chicken, Mouse, Clone: TD77, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma 051144
Description
Detect Talin using this Anti-Talin Antibody, C-terminus, clone TD77 validated for use in western blot, immunohistochemistry (paraffin).Specifications
Talin | |
Monoclonal | |
Unconjugated | |
Q9Y490 | |
Mouse | |
Purified from Protein A Sepharose™ chromatography | |
Primary | |
2-8°C in for up to 12 months. | |
IgG1 |
Immunohistochemistry (Paraffin), Western Blot | |
TD77 | |
Purified immunoglobulin from Protein A Sepharose™ chromatography. Liquid in 0.02M PBS pH 7.6, 0.25MNaCl containing 0.1% sodium azide. | |
ILWEQ , KIAA1027, TLN | |
Human platelet protein | |
100 μg | |
Chicken, Human, Mouse | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only