Learn More
MilliporeSigma™ anti-TANK, Polyclonal Available on GSA/VA Contract for Federal Government customers only.
Rabbit Polyclonal Antibody
Manufacturer:  MilliporeSigma™ ST1608100UG
Catalog No. ST1608100UG
Add to cart
Description
Description
Specifically detects TANK Clone: in Human samples, and it is validated for Immuno Blotting
The TRAF (tumor necrosis factor receptor-associated factor) family of proteins associate with and transduce signals from members of the tumor necrosis factor receptor superfamily. The protein encoded by this gene is found in the cytoplasm and can bind to TRAF1, TRAF2, or TRAF3, thereby inhibiting TRAF function by sequestering the TRAFs in a latent state in the cytoplasm. For example, the protein encoded by this gene can block TRAF2 binding to LMP1, the Epstein-Barr virus transforming protein, and inhibit LMP1-mediated NF-κB activation. Two transcript variants encoding different isoforms have been found for this gene.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
TANK
Unconjugated
Purified
Full-length, human TANK (aa 1-119)
100μg
−20°C
Polyclonal
Human
Immuno Blotting, Immunoblot
In PBS, pH 7.2.
Rabbit
IgG
RUO
Primary
Recognizes the ∽43-45kDa TANK protein in human liver tissue.