Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TAP1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA131487

Catalog No. PA131487


Add to cart

Description

Description

PA1-31487 detects TAP I from human samples. PA1-31487 has been successfully used in flow cytometry, immunoprecipitation, Western blot applications. The PA1-31487 immunogen is: Transporter Associated with Antigen Processing (TAP-I ) peptide corresponding to the C-terminus of the human protein conjugated to Keyhole Limpet Hemocyanin (KLH). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

The membrane-associated protein encoded by this gene is a member of the superfamily of ATP-binding cassette (ABC) transporters. ABC proteins transport various molecules across extra- and intra-cellular membranes. ABC genes are divided into seven distinct subfamilies (ABC1, MDR/TAP, MRP, ALD, OABP, GCN20, White). This protein is a member of the MDR/TAP subfamily. Members of the MDR/TAP subfamily are involved in multidrug resistance. The protein encoded by this gene is involved in the pumping of degraded cytosolic peptides across the endoplasmic reticulum into the membrane-bound compartment where class I molecules assemble. Mutations in this gene may be associated with ankylosing spondylitis, insulin-dependent diabetes mellitus, and celiac disease.
Specifications

Specifications

TAP1
Polyclonal
Unconjugated
TAP1
Liquid
TAP1
Transporter Associated with Antigen Processing (TAP-I ) peptide corresponding to the C-terminus of the human protein conjugated to Keyhole Limpet Hemocyanin (KLH).
IgG fraction
RUO
Antibody
Polyclonal
Human
ELISA, Flow Cytometry, Immunoprecipitation, Western Blot
1 mg/mL
0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2
Q03518
DAAP-57C1.5, ABC17, ABCB2, APT1, D6S114E, FLJ26666, FLJ41500, PSF1, RING4, TAP1*0102N, TAP1N
Rabbit
IgG
100 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
6890
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit