Learn More
TAP1 Mouse anti-Human, Clone: 3D4, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA520071
Catalog No. PIMA520071
Add to cart
Description
Description
Peptide Sequence: GSETRRLSLF LVLVVLSSLG EMAIPFFTGR LTDWILQDGS ADTFTRNLTL MSILTIASAV LEFVGDGIYN NTMGHVHSHL QGEVFGAVLR QETEFFQQNQ TGNIMSRVTE
The membrane-associated protein encoded by this gene is a member of the superfamily of ATP-binding cassette (ABC) transporters. ABC proteins transport various molecules across extra- and intra-cellular membranes. ABC genes are divided into seven distinct subfamilies (ABC1, MDR/TAP, MRP, ALD, OABP, GCN20, White). This protein is a member of the MDR/TAP subfamily. Members of the MDR/TAP subfamily are involved in multidrug resistance. The protein encoded by this gene is involved in the pumping of degraded cytosolic peptides across the endoplasmic reticulum into the membrane-bound compartment where class I molecules assemble. Mutations in this gene may be associated with ankylosing spondylitis, insulin-dependent diabetes mellitus, and celiac disease.
Specifications
Specifications
TAP1
Monoclonal
1 mg/mL
PBS with no preservative; pH 7.4
Q03518
ABP-278, ABP-280, AOI, FH1, FLN-B, FLN1L, LRS1, SCT, TABP, TAP, ABP-280 homolog, Larsen syndrome 1 (autosomal domit), actin binding protein 278, actin-binding-like protein, beta-filamin, filamin homolog 1, filamin-3, filamin-B, thyroid autoantigen
Mouse
IgG3, kappa
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
6890
ELISA, Immunocytochemistry, Immunofluorescence, Western Blot
3D4
Unconjugated
TAP1
Liquid
TAP1
TAP1 (AAH14081, 241 a.a. ∽ 350 a.a) partial recombit protein with GST tag. MW of the GST tag alone is 26 KDa.
Affinity chromatography
RUO
Antibody
Monoclonal
Human