Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TAPP1 Rat anti-Human, Clone: 376409, Invitrogen™

Rat Monoclonal Antibody

Manufacturer:  Invitrogen MA524117

Catalog No. PIMA524117


Add to cart

Description

Description

Reconstitute at 0.5 mg/mL in sterile PBS.

Specifications

Specifications

TAPP1
Monoclonal
0.5 mg/mL
PBS with 5% trehalose and No Preservative
Q9HB21
PLEKHA1, family A (phosphoinositide bindingspecific) member 1, PH domain-containing family A member 1, pleckstrin homology domain containing, family A (phosphoinositide bindingspecific) member 1
Rat
IgG2A
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
59338
Western Blot
376409
Unconjugated
PLEKHA1
Lyophilized
PLEKHA1
E. coli-derived recombit human TAPP1 Pro2-Val404
Protein A/G
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit