Learn More
TARS Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier:  Novus Biologicals NBP220567
Catalog No. NBP220567
Add to cart
Description
Description
TARS Polyclonal antibody specifically detects TARS in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin).
Specifications
Specifications
TARS
Polyclonal
Unconjugated
0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal
cytoplasmic, EC 6.1.1.3, MGC9344, Threonine--tRNA ligase, threonyl-tRNA synthetase, threonyl-tRNA synthetase, cytoplasmic
Rabbit
83 kDa
0.1 mL
metabolism
6897
Human, Mouse
IgG
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
0.27mg/mL
Western Blot 1:500-1:3000, Immunohistochemistry 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000
P26639
TARS
Recombinant protein encompassing a sequence within the center region of human ThrRS. The exact sequence is proprietary.
Affinity Purified
RUO
Primary
Human, Mouse. Species reactivity based on sequence identity: Chicken (91%), Rat (98%), Xenopus laevis (93%), Zebrafish (93%), Bovine (96%), Xenopus tropicalis (93%).
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.