Learn More
TAS2R9, Rabbit, Monoclonal Antibody, Abnova™
Rabbit monoclonal antibody raised against a human TAS2R9 peptide using ARM Technology.
Supplier: Abnova Corporation H00050835K
Includes: Up to three rabbit IgG clones of 1mg each will be delivered to customer.
Description
This gene product belongs to the family of candidate taste receptors that are members of the G-protein-coupled receptor superfamily. These proteins are specifically expressed in the taste receptor cells of the tongue and palate epithelia. They are organized in the genome in clusters and are genetically linked to loci that influence bitter perception in mice and humans. In functional expression studies, they respond to bitter tastants. This gene maps to the taste receptor gene cluster on chromosome 12p13. [provided by RefSeq
Library Construction: Nonfusion antibody library from rabbit spleen (ARM Technology)Expression: Overexpression vector and transfection into 293H cell line
Order Info
- Customer may provide cell or tissue lysate for antibody screening.
- Rabbit monoclonal antibody generated by ARM technology is amenable to antibody engineering including F(ab)2, IgG, scFv and different Fc and non-Fc conjugates per customer request.
Specifications
TAS2R9 | |
Monoclonal | |
Unconjugated | |
In 1X PBS, pH 7.4 | |
TAS2R9 | |
TAS2R9 | |
A synthetic peptide of human TAS2R9 | |
RUO | |
50835 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
ELISA, Western Blot | |
1mg | |
taste receptor, type 2, member 9 | |
TAS2R9 | |
T2R9, TRB6 | |
Rabbit | |
1 mg x up to 3 Abs | |
Primary | |
Human | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.