Learn More
TAS1R2 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen OSR00180W
Catalog No. OSR00180W
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Glycerol (1:1) may be added for additional stability. Reconstitute in 100 µL of sterile water.
Putative taste receptor. TAS1R2/TAS1R3 recognizes diverse natural and synthetic sweeteners.
Specifications
Specifications
TAS1R2
Polyclonal
whole serum with no preservative
Q8TE23
Taste receptor type 1 member 2 (precursor), sweet taste receptor T1R2, G-protein coupled receptor 71, TAS1R2, TR2, T1R2, GPR71, TS1R2
Rabbit
IgG
RUO
Antibody
Polyclonal
Human
Immunohistochemistry, Western Blot
Unconjugated
TAS1R2
Lyophilized
TAS1R2
A synthetic peptide from c-terminal region of human taste receptor, type 1, member 2
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
80834