Learn More
TAS2R43 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen OSR00167W
Catalog No. OSR00167W
Add to cart
Description
Description
Reconstitute with 100 µL of distilled water. Centrifuge to remove any insoluble material.
Specifications
Specifications
TAS2R43
Polyclonal
whole serum with no preservative
P59537
Taste receptor type 2 member 43, T2R43, T2R52, TAS2R43
Rabbit
IgG
RUO
Antibody
Polyclonal
Human
Immunohistochemistry, Western Blot
Unconjugated
TAS2R43
Lyophilized
TAS2R43
Synthetic peptide from human taste receptor, type 2, member 43.
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
259289