Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rat Tau Mouse anti-Rat, Unlabeled, Clone: 15/TAU, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 610672

Catalog No. BDB610672


Add to cart

Description

Description

In addition to tubulin, microtubules have other components named Microtubule-associated proteins (MAPs), which include the high molecular MAPs (200-400 kDa) and the Tau proteins (55-70 kDa). These proteins co-purify with the microtubules during repeated cycles of microtubule assembly and disassembly. Both MAPs and Tau proteins extend from the side of the microtubules. They form cross-bridges in neurons and bind tubulin by their C-terminal region. The heterogeneity in the molecular weight of Tau proteins is due to post-translational modifications and different numbers of tandem repeats in their C-terminal domain. Tau isoforms are selectively expressed in unique subsets of neurons and regulated during development. The highly phosphorylated Tau proteins have also been detected in the neurofibrillary tangles of Alzheimer's patients.

Specifications

Specifications

Rat Tau
15/TAU
Unconjugated
Affinity Purified
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
50μg
Cell Biology
Primary
Rat
Western Blot
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Murine
IgG2b, κ
RUO
Store undiluted at -20°C.
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit