Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TAX1BP1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
$409.50
Specifications
Antigen | TAX1BP1 |
---|---|
Concentration | 0.5 mg/mL |
Dilution | Western Blot 2 ug/ml |
Applications | Western Blot |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB600975
|
Novus Biologicals
NB600975 |
0.1 mg |
Each of 1 for $409.50
|
|
|||||
Description
TAX1BP1 Polyclonal specifically detects TAX1BP1 in Human samples. It is validated for Western Blot.Specifications
TAX1BP1 | |
Western Blot 2 ug/ml | |
Polyclonal | |
Purified | |
RUO | |
Human | |
NP_001073333 | |
8887 | |
This antibody was developed against amino acids 718-735 (CSEQFPPDYDQQVFERHV) of human TRAF6BP (TAX1BP). The sequence used is 94% homologous in mouse. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
0.5 mg/mL | |
Western Blot | |
Unconjugated | |
Rabbit | |
Neuroscience | |
PBS containing 0.05% BSA with 0.05% Sodium Azide | |
T6BP, Tax1 (human T-cell leukemia virus type I) binding protein 1, tax1-binding protein 1, TRAF6-binding protein, TXBP151CALCOCO3 | |
TAX1BP1 | |
IgG | |
Protein G purified | |
There are multiple isoforms of TRAF6BP ranging from 590 amino acids to 858 amino acids. Researchers are encouraged to titer antibody for their specific applications. |
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title