Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TBC1D14 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524473

Catalog No. PIPA524473


Add to cart

Description

Description

May act as a GTPase-activating protein for Rab family protein(s).
Specifications

Specifications

TBC1D14
Polyclonal
PBS with 0.09% sodium azide
Q9P2M4, Q8CGA2
TBC1 domain family member 14
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
100855, 57533
Immunohistochemistry, Western Blot
Unconjugated
TBC1D14
Liquid
TBC1D14
KLH conjugated synthetic peptide between 259-286 amino acids from the central region of human TBC14
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit