Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TBCC Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA521710

Catalog No. PIPA521710


Add to cart

Description

Description

PA5-21710 targets TBCC in WB applications and shows reactivity with Human and mouse samples. The PA5-21710 immunogen is recombit protein fragment contain a sequence corresponding to a region within amino acids 52 and 306 of TBCC. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

Specifications

Specifications

TBCC
Polyclonal
Unconjugated
TBCC
Liquid
TBCC
Recombit protein fragment contain a sequence corresponding to a region within amino acids 52 and 306 of TBCC
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Immunohistochemistry (Paraffin), Western Blot
0.82 mg/mL
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Q15814, Q8VCN9
CFC, tubulin-folding cofactor C, tubulin-specific chaperone C
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
6903, 72726
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit