Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TBCCD1 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524074

Catalog No. PIPA524074


Add to cart

Description

Description

The function of TBCCD1 remains unknown.
Specifications

Specifications

TBCCD1
Polyclonal
PBS with 0.09% sodium azide
Q9NVR7, Q640P7
TBCC domain-containing protein 1
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
55171, 70573
Western Blot
Unconjugated
TBCCD1
Liquid
TBCCD1
KLH conjugated synthetic peptide between 491-520 amino acids from the C-terminal region of human TBCCD1
Protein A, Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit