Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TBC1D7 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA526899

Catalog No. PIPA526899


Add to cart

Description

Description

TBC1D7 belongs to the family of proteins sharing a 180- to 200-amino acid TBC domain presumed to have a role in regulating cell growth and differentiation. These proteins share significant homology with TRE2.
Specifications

Specifications

TBC1D7
Polyclonal
PBS with 0.09% sodium azide
Q9P0N9, Q9D0K0
PIG51, TBC7, dJ257A7.3, TBC1 domain family member 7, cell migration-inducing protein 23
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
51256, 67046
Western Blot
Unconjugated
TBC1D7
Liquid
TBC1D7
KLH conjugated synthetic peptide between 150-179 amino acids from the central region of human TBCD7
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit