Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TBCE Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA526674

Catalog No. PIPA526674


Add to cart

Description

Description

Cofactor E is one of four proteins (cofactors A, D, E, and C) involved in the pathway leading to correctly folded beta-tubulin from folding intermediates. Cofactors A and D are believed to play a role in capturing and stabilizing beta-tubulin intermediates in a quasi-native confirmation. Cofactor E binds to the cofactor D/beta-tubulin complex; interaction with cofactor C then causes the release of beta-tubulin polypeptides that are committed to the native state.
Specifications

Specifications

TBCE
Polyclonal
PBS with 0.09% sodium azide
Q15813
HRD, KCS, KCS1, pac2, Kenny-Caffey syndrome, tubulin-folding cofactor E, tubulin-specific chaperone E
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
6905
Western Blot
Unconjugated
TBCE
Liquid
TBCE
KLH conjugated synthetic peptide between 364-392 amino acids from the central region of human TBCE
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit