Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TBK1 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA515053
Description
This antibody is predicted to react with mouse and Xenopus based on sequence homology.
The NF-kappa-B (NFKB) complex of proteins is inhibited by I-kappa-B (IKB) proteins, which inactivate NFKB by trapping it in the cytoplasm. Phosphorylation of serine residues on the IKB proteins by IKB kinases marks them for destruction via the ubiquitination pathway, thereby allowing activation and nuclear translocation of the NFKB complex. TKB is similar to IKB kinases and can mediate NFKB activation in response to certain growth factors. The protein can form a complex with the IKB protein TANK and TRAF2 and release the NFKB inhibition caused by TANK.Specifications
TBK1 | |
Polyclonal | |
TBK1 | |
NAK, Serine/threonine-protein kinase TBK1, TANK-binding kinase 1, T2K, NF-kappa-B-activating kinase, TBK1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
29110 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry, Western Blot | |
Unconjugated | |
Q9UHD2 | |
TBK1 | |
KLH conjugated synthetic peptide between 150-181 amino acids from human TBK | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction