Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TBRG1 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA513390

Catalog No. PIPA513390

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

TBRG1 acts as a growth inhibitor. It can activate TP53/p53, cause G1 arrest and collaborate with CDKN2A to restrict proliferation, but it does not require either protein to inhibit DNA synthesis. TBRG1 redistributes CDKN2A into the nucleoplasm, and is involved in maintaining chromosomal stability.
Specifications

Specifications

TBRG1
Polyclonal
PBS with 0.09% sodium azide
Q3YBR2, Q3UB74
Transforming growth factor beta regulator 1; Nuclear interactor of ARF and Mdm2; TBRG1; NIAM
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
21376, 84897
Western Blot
Unconjugated
TBRG1
Liquid
TBRG1
KLH conjugated synthetic peptide between 6-36 amino acids from the N-terminal region of human TBRG1
Ammonium sulfate precipitation, Size-exclusion - Dialysis
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit