Learn More
TBX19 Rabbit anti-Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA1803
Description
Tpit (TBX19, T-Box protein 19) is a highly cell-restricted transcription factor belonging to the T-Box family. Tpit is required for pro-opeiomelanocortin (POMC) gene expression and differentiation of corticotroph and melanotroph cells. Tpit gene mutations have been shown to be directly correlated to pituitary adrenocorticotropin (ACTH) deficiencies. Tpit functions as a transcriptional activator by recruiting the p160 family of coactivators.
PA1-803 detects endogenous Tpit from mouse AtT-20 cell lysates.PA1-803 has been successfully used in Western blot procedures. By Western blot, this antibody detects an ∽48kDa protein representing Tpit from AtT-20 cells. PA1-803 immunizing protein, bacterially produced, corresponds to the mouse C-terminal region of Tpit.Specifications
TBX19 | |
Polyclonal | |
Tbx19 | |
TBX19, T-Box protein 19 | |
Rabbit | |
Ammonium sulfate precipitation | |
RUO | |
83993 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
Unconjugated | |
Q99ME7 | |
TBX19 | |
Mouse recombit protein from C-terminal region Tpit. | |
100 μL | |
Primary | |
Mouse | |
Antibody | |
IgG |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.