Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-TBX2, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP133551

Catalog No. NBP133551


Add to cart

Description

Description

TBX2 Polyclonal antibody specifically detects TBX2 in Human samples. It is validated for Western Blot.
Specifications

Specifications

TBX2
1.0mg/mL
Western Blot 1:500-1:3000
Affinity Purified
TBX2
Recombinant protein encompassing a sequence within the center region of human TBX2. The exact sequence is proprietary.
75 kDa
100 ul
Cancer
Primary
6909
Human
Western Blot
Unconjugated
0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal
FLJ10169, T-box 2, T-box protein 2, T-box transcription factor TBX2
Rabbit
IgG
Immunogen affinity purified
RUO
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Polyclonal
Tested in human. Not yet tested in any other species.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only