Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-TCEA1, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP126625

Catalog No. NBP126625


Add to cart

Description

Description

TCEA1 Polyclonal antibody specifically detects TCEA1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunoprecipitation (Negative).
Specifications

Specifications

TCEA1
0.2 mg/mL
Western Blot 1:2000-1:10000, Immunohistochemistry 1:200- 1:1000, Immunoprecipitation, Immunohistochemistry-Paraffin 1:200- 1:1000
P23193
GTF2Stranscription elongation factor A protein 1, SII, TF2STCEA, TFIISTranscription elongation factor TFIIS.o, transcription elongation factor A (SII), 1, Transcription elongation factor S-II protein 1
Rabbit
IgG
0.1 ml
Store at 4C. Do not freeze.
Polyclonal
Human, Mouse. Based on 100% sequence identity, this antibody is predicted to react with Rat, Bovine, Dog, Horse, Rabbit, Guinea pig_10141, Pig, Panda, Orangutan, Rhesus Monkey, Gorilla, Chimpanzee, White-tuftedear marmoset, Chinese hamster, Naked mole rat, African elephant, and Gray short-tailed opossum.
Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Unconjugated
TBS and 0.1% BSA with 0.09% Sodium Azide
Affinity Purified
TCEA1
The immunogen recognized by this antibody maps to a region between residue 200 and 250 of human transcription elongation factor S-II (transcription elongation factor A (SII), 1) using the numbering given in entry NP_006747.1 (GeneID 6917).
Immunogen affinity purified
RUO
Primary
6917
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only