Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-TCEAL1, Polyclonal, Novus Biologicals™

Goat Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP136995

Catalog No. NBP136995


Add to cart

Description

Description

TCEAL1 Polyclonal antibody specifically detects TCEAL1 in Human samples. It is validated for Peptide ELISA.
Specifications

Specifications

TCEAL1
0.5 mg/mL
Peptide ELISA Detection limit 1:8000
NP_004771.2
Nuclear phosphoprotein p21/SIIR, p21, pp21, SIIRTCEA-like protein 1, transcription elongation factor A (SII)-like 1, transcription elongation factor A protein-like 1, Transcription elongation factor S-II protein-like 1
Goat
IgG
0.1 mg
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
Reported variants represent identical protein: NP_001006641.1, NP_001006640.1, NP_004771.2.
ELISA
Unconjugated
0.5 mg/ml Tris (pH 7.3) and 0.5% BSA with 0.02% Sodium Azide
Affinity Purified
TCEAL1
Peptide with sequence DKPRKENEEEPQ-C corresponding to N-Terminus according to NP_004771.2.
Immunogen affinity purified
RUO
Primary
9338
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only