Learn More
TCF-1 Mouse anti-Human, Clone: 7H3, MilliporeSigma™
Mouse Monoclonal Antibody
Manufacturer:  MilliporeSigma 05510
Catalog No. 05-510-MI
Add to cart
Description
Description
Detect TCF-1 with Anti-TCF-1 Antibody, clone 7H3 (Mouse Monoclonal Antibody), that has been shown to work in immunocytochemistry, immunohistochemistry and western blotting.
Specifications
Specifications
TCF-1
7H3
Protein G Purified immunoglobulin in Immunoaffinity Purified immunoglobulin in 0.2M Tris-glycine, pH 7.5, 0.15M NaCl and 0.05% sodium azide
Purified
Murine
IgG
100μg
Primary
Human
Immunocytochemistry, Immunohistochemistry, Western Blot
Unconjugated
NM_003202
TCF1, MODY3, TCF-1, HNF1, HNF-1A, HNF1A, LFB1, HNF1a
Maltose binding protein (MBP) fusion protein corresponding to residues 102-269 of human TCF-1A splice variant. The epitope maps to a region just N-terminal to the High Mobility Group (HMG) box
Protein G purfied
−20°C in undiluted aliquots for up to 24 months. Avoid repeated freeze/thaw cycles.
Monoclonal