Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TCF19 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA521744
Description
Recommended positive controls: 293T. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
TCF19 Polyclonal antibody specifically detects TCF19 in Human samples. It is validated for Western Blot.Specifications
TCF19 | |
Polyclonal | |
Unconjugated | |
TCF19 | |
5730403J10Rik; AW495861; DAMA-213L4.5; SC 1; SC1; SC1 protein homolog; SC1(TCF19)-6; SC1(TCF19)-7; SC1-1; SCI(TCF19)-4; TCF 19; TCF19; TCF-19; Transcription factor 19; transcription factor 19-like protein; Transcription factor SC1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
6941 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.96 mg/mL | |
PBS with 10% glycerol and 0.01% thimerosal; pH 7 | |
Q9Y242 | |
TCF19 | |
Synthetic peptide contain a sequence corresponding to a region within amino acids 296 and 309 (Q9Y242) of TCF19 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction