Learn More
TCF7L2 Mouse anti-Human, Mouse, Clone: 6H5.3, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA112483
Catalog No. 01-674-331
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
A suggested positive control for this product is COS-1 or HCT116 cells.
TCF7L2, or TCF4, is a high mobility group (HMG) box-containing transcription factor that plays a key role in the Wnt signaling pathway. TCF7L2 has been implicated in glucose homeostasis through the regulation of pro-glucagon gene expression, which encodes glucagon-like peptide 1 (GLP-1). Genetic variants of TCF7L2 have been linked to type 2 diabetes, and it has specifically been shown to have an important role in the regulation of both beta-cell survival and function. Several splicing isoforms for TCF7L2 have been shown to be differentially expressed in tissues during cancer progression, and gene targeting studies indicate that TCF7L2 is required to maintain the crypt stem cells of the small intestine.
Specifications
Specifications
TCF7L2
Monoclonal
1 mg/mL
0.7M tris glycine with 0.105M NaCl, 30% glycerol and 0.05% sodium azide; pH 7.4
Q9NQB0, Q924A0
TCF7L2
Mouse
IgG2a
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
21416, 6934
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
6H5.3
Unconjugated
TCF7L2
Liquid
TCF7L2
Recombit protein corresponding to residues 31-331 of human TCF-4.
Protein A/G
RUO
Antibody
Monoclonal
Human, Mouse