Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TCF7 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA511794

Catalog No. PIPA511794

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

The T cell specific transcription factor TCF7 activates genes involved in immune regulation and thymocyte differentiation, and is a candidate locus for genetic susceptibility to type 1 diabetes.
Specifications

Specifications

TCF7
Polyclonal
Unconjugated
TCF7
Liquid
TCF7
KLH conjugated synthetic peptide between 3-36 amino acids from the N-terminal region of human TCF7
Ammonium sulfate precipitation, Size-exclusion - Dialysis
RUO
Antibody
Polyclonal
Human, Mouse
Western Blot
2.0 mg/mL
PBS with 0.09% sodium azide
P36402, Q00417
Transcription factor 7; T-cell specific; HMG-box; TCF7;
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
21414, 6932
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit