Learn More
anti-TCOF1, Polyclonal, Abnova
Mouse Polyclonal
Supplier: Abnova Corporation H00006949B01
Description
This gene encodes a nucleolar protein with a LIS1 homology domain. The protein is involved in ribosomal DNA gene transcription through its interaction with upstream binding factor (UBF). Mutations in this gene have been associated with Treacher Collins syndrome, a disorder which includes abnormal craniofacial development. Alternate transcriptional splice variants encoding different isoforms have been found for this gene, but only three of them have been characterized to date.
Detection Antibody, Western Blotting
Specifications
Treacher Collins-Franceschetti syndrome 1 | |
Unconjugated | |
TCOF1 (NP_001008657.1, 1 a.a. ∽958 a.a) full-length human protein. | |
RUO | |
Primary | |
Serum |
Polyclonal | |
Mouse | |
50 μL | |
Yes | |
Human |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.