Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TCP-1 eta Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA527393

Catalog No. PIPA527393


Add to cart

Description

Description

PA5-27393 targets TCP1 eta in WB applications and shows reactivity with Human samples. The PA5-27393 immunogen is recombit fragment corresponding to a region within amino acids 361 and 530 of Human TCP1 eta. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

This gene encodes a molecular chaperone that is a member of the chaperonin containing TCP1 complex (CCT), also known as the TCP1 ring complex (TRiC). This complex consists of two identical stacked rings, each containing eight different proteins. Unfolded polypeptides enter the central cavity of the complex and are folded in an ATP-dependent manner. The complex folds various proteins, including actin and tubulin. Alternate transcriptional splice variants encoding different isoforms have been found for this gene, but only two of them have been characterized to date.
Specifications

Specifications

TCP-1 eta
Polyclonal
Unconjugated
CCT7
Liquid
CCT7
Recombit fragment corresponding to a region within amino acids 361 and 530 of Human TCP1 eta
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Western Blot
0.2 mg/mL
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
Q99832
CCTETA, CCTH, NIP7-1, TCP1ETA, CCT-eta, HIV-1 Nef interacting protein, HIV-1 Nef-interacting protein, T-complex protein 1 subunit eta, TCP-1-eta, chaperonin containing t-complex polypeptide 1, eta subunit
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
10574
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit