Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TCP1 Goat anti-Human, Polyclonal, Invitrogen™

Goat Polyclonal Antibody

Manufacturer:  Invitrogen PA518251

Catalog No. PIPA518251


Add to cart

Description

Description

The protein encoded by this gene is a molecular chaperone that is a member of the chaperonin containing TCP1 complex , also known as the TCP1 ring complex . This complex consists of two identical stacked rings, each containing eight different proteins. Unfolded polypeptides enter the central cavity of the complex and are folded in an ATP-dependent manner. The complex folds various proteins, including actin and tubulin. Alternate transcriptional splice variants of this gene, encoding different isoforms, have been characterized. In addition, three pseudogenes that appear to be derived from this gene have been found.
Specifications

Specifications

TCP1
Polyclonal
Unconjugated
TCP1
Liquid
TCP1
Synthetic peptide sequence (SYEDAVHSGALND) corresponding to the C-terminus amino acids of TCP1
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunohistochemistry, Western Blot
0.5 mg/mL
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
P17987
TCP1; t-complex 1; CCT-alpha; CCT1; CCTa; D6S230E; TCP-1-alpha; T-complex protein 1; T-complex protein 1, alpha subunit; tailless complex polypeptide 1
Goat
IgG
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
6950
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit