Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-TCR alpha, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP145883

Catalog No. NBP145883


Add to cart

Description

Description

TCR alpha Polyclonal antibody specifically detects TCR alpha in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation.
Specifications

Specifications

TCR alpha
0.5 mg/ml
Western Blot 0.5 - 4 ug/mL, Immunohistochemistry 15 - 25 ug/mL, Immunoprecipitation 10 - 20 ug/mL
Affinity Purified
TRA
Synthetic peptide surrounding amino acid 138 of human TCR&alpha.
Affinity purified
RUO
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Polyclonal
Human, Mouse, Rat
Immunohistochemistry, Immunoprecipitation, Western Blot
Unconjugated
PBS (pH 7.2), 30% glycerol, 0.5% BSA, 5 mM EDTA with 0.03% Proclin
C), D, FLJ22602, J, MGC117436, MGC22624, MGC23964, MGC71411, T cell receptor alpha locus, T-cell antigen receptor, alpha polypeptide, T-cell receptor, alpha (V, TCRA, TCRD, TRA
Rabbit
IgG
0.1 mg
Adaptive Immunity, Cell Biology, Immunology, Protein Kinase, Signal Transduction, Tyrosine Kinases
Primary
6955
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only