Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-TCR beta chain, APC, Clone: H57-597,

Armenian Hamster Monoclonal Antibody

$153.50 - $237.00

Specifications

Antigen TCR beta chain
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Format Purified
Formulation Purified Armenian Hamster monoclonal containing 10mM NaH2PO4, 150mM NaCl, 0.09% NaN3, 0.1% gelatin, pH 7.2.
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MABF1452MI
View Documents
MilliporeSigma™
MABF1452
25μg
Each for $153.50
Add to cart
 
MABF1453MI
View Documents
MilliporeSigma™
MABF1453
100μg
Each for $237.00
Add to cart
 
Description

Description

Specifically detects TCR beta chain Clone: H57-597 in Mouse samples, and it is validated for Flow Cytometry

The H57-597 antibody is specific for the beta chain of the mouse T cell Receptor (TCR). This cell surface protein combines with a second protein chain (alpha chain) to form the alpha-beta TCR that is expressed by NK1.1+ thymocytes, NKT cells, and the majority of peripheral T cells. A small number of T cells may express an alternative heteromer of gamma and delta protein chains, known as the gamma-delta TCR. These receptors participate in a complex with CD3, and with the co-receptors CD4 or CD8, to recognize and respond to antigens bound to MHC molecules on antigen-presenting cells. Such interactions promote T cell receptor signaling (T cell activation) and can result in a number of cellular responses including proliferation, differentiation, production of cytokines or activation-induced cell death. The H57-597 antibody is used as a phenotypic marker for T cells expressing the alpha-beta TCR. It is also widely used to cross-link surface TCR and thereby mimic TCR-mediated cell activation or induction of apoptosis. The antibody does not cross-react with cells expressing the gamma-delta TCR.
Specifications

Specifications

TCR beta chain
Monoclonal
Purified Armenian Hamster monoclonal containing 10mM NaH2PO4, 150mM NaCl, 0.09% NaN3, 0.1% gelatin, pH 7.2.
Flow Cytometry
IgG
P0C6B2
2°C to 8°C for one year from date of shipment protected from light; Do not freeze
Inflammation, Immunology
Primary
Purified
H57-597
Armenian Hamster
Proprietary immunogen
Ptcra, pT-alpha-TCR
RUO
Purified by size-exclusion chromatography
SDS
Product Certifications

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit