Learn More
TCR αβ Mouse anti-Human, Brilliant Violet 786, Clone: T10B9.1A-31 (also known as T10B9), BD
Mouse Monoclonal Antibody
Manufacturer:  BD Biosciences 563825
Catalog No. BDB563825
Add to cart
Description
Description
The T10B9.1A-31 monoclonal antibody specifically binds to a monomorphic determinant on the αβ T-cell receptor expressed on greater than 95% of normal peripheral blood CD3+ T cells. The αβ TCR recognizes a peptide bound to MHC leading to T-cell activation. The T10B9.1A-31 antibody induces T-cell activation in the immobilized form and is useful in flow cytometry for studying αβ + T-cell populations.
Specifications
Specifications
TCR αβ
T10B9.1A-31 (also known as T10B9)
αβ TCR; TCR alpha-beta
Affinity Purified
Human Peripheral Blood T Cells
100 Tests
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal
Flow Cytometry
Brilliant Violet 786
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Mouse
IgM, κ
RUO
Primary
Human