Learn More
TDRD7 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA524462
Catalog No. PIPA524462
Add to cart
Description
Description
This antibody is predicted to react with bovine and rat based on sequence homology.
TDRD7, also known as Tudor domain containing protein 7, is a component of chromatoid bodies in spermatids (named processing bodies or P-bodies in somatic cells). It is found in a mRNP complex together with TDRD1, TDRD6 and DDX4. It is also found in a complex with CABLES1, PCTK2 and PIWIL1. TDRD7 has been shown to interact with histone H3 tri-methylated at K9 in vitro.
Specifications
Specifications
TDRD7
Polyclonal
Unconjugated
TDRD7
Liquid
TDRD7
KLH conjugated synthetic peptide between 807-835 amino acids from the C-terminal region of human TDRD7
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
0.5 mg/mL
PBS with 0.09% sodium azide
Q8NHU6, Q8K1H1
CATC4, PCTAIRE2BP, RP11-508D10.1, TRAP, PCTAIRE2-binding protein, tudor domain-containing protein 7, tudor repeat associator with PCTAIRE 2
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
100121, 23424